5-Fluoro-2-(trifluoromethoxy)benzamide

Lieferant: Apollo Scientific
Achtung

PC302618-1G PC302618-5G
APOSPC302618-1GEA 0 EUR
APOSPC302618-1G APOSPC302618-5G
5-Fluoro-2-(trifluoromethoxy)benzamide
5-Fluoro-2-(trifluoromethoxy)benzamide
Formel: C₈H₅F₄NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11519335
CAS-Nummer: 1092460-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR