5-Fluoro-2-methylcinnamic acid 97%

Lieferant: Apollo Scientific

PC302624-5G PC302624-1G
APOSPC302624-5GEA 159 EUR
APOSPC302624-5G APOSPC302624-1G
5-Fluoro-2-methylcinnamic acid 97%
5-Fluoro-2-methylcinnamic acid
Formel: C₁₀H₉FO₂
Molecular Weight: 180,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06205343
CAS-Nummer: 773129-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR