2,3-Difluoro-4-methoxybenzyl alcohol 98%
Lieferant: Apollo Scientific
Synonyme:
(2,3-Difluoro-4-methoxyphenyl)methanol
PC302658-5G
PC302658-1G
PC302658-25G
APOSPC302658-5GEA
178
EUR
APOSPC302658-5G
APOSPC302658-1G
APOSPC302658-25G
2,3-Difluoro-4-methoxybenzyl alcohol 98%
2,3-Difluoro-4-methoxybenzyl alcohol
Formel:
C₈H₈F₂O₂ Molecular Weight: 174.145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115984 CAS-Nummer: 886500-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...