3-[2-Methyl-5-(trifluoromethyl)phenyl]propionic acid 98%

Lieferant: Apollo Scientific

PC302706-1G PC302706-5G PC302706-10G
APOSPC302706-1GEA 203 EUR
APOSPC302706-1G APOSPC302706-5G APOSPC302706-10G
3-[2-Methyl-5-(trifluoromethyl)phenyl]propionic acid 98%
3-[2-Methyl-5-(trifluoromethyl)phenyl]propionic acid
Formel: C₁₁H₁₁F₃O₂
Molecular Weight: 232.199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832289
CAS-Nummer: 1017778-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR