4-Chloro-3,5-difluorobenzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC302742-5G PC302742-1G
APOSPC302742-5GEA 522 EUR
APOSPC302742-5G APOSPC302742-1G
4-Chloro-3,5-difluorobenzoyl chloride 97%
4-Chloro-3,5-difluorobenzoyl chloride
Formel: C₇H₂Cl₂F₂O
Molecular Weight: 210,993 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415577
CAS-Nummer: 1261799-38-5
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR