3-Chloro-4-ethoxy-5-fluorobenzamide
Lieferant: Apollo Scientific
PC302770-10G
PC302770-5G
APOSPC302770-10GEA
351
EUR
APOSPC302770-10G
APOSPC302770-5G
3-Chloro-4-ethoxy-5-fluorobenzamide
3-Chloro-4-ethoxy-5-fluorobenzamide
Formel:
C₉H₉ClFNO₂ Molecular Weight: 217.625 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832338 CAS-Nummer: 1017778-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...