2-Methyl-5-(trifluoromethyl)benzylamine
Lieferant: Apollo Scientific
|
Gefahr
|
PC302821-10G
PC302821-5G
PC302821-1G
APOSPC302821-10GEA
361
EUR
APOSPC302821-10G
APOSPC302821-5G
APOSPC302821-1G
2-Methyl-5-(trifluoromethyl)benzylamine
2-Methyl-5-(trifluoromethyl)benzylamine
Formel:
C₉H₁₀F₃N Molecular Weight: 189.178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06213552 CAS-Nummer: 771572-44-2 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...