3,5-Difluoro-2-methoxybenzoyl chloride

Lieferant: Apollo Scientific

PC302823-10G PC302823-5G
APOSPC302823-10GEA 282 EUR
APOSPC302823-10G APOSPC302823-5G
3,5-Difluoro-2-methoxybenzoyl chloride
3,5-Difluoro-2-methoxybenzoyl chloride
Formel: C₈H₅ClF₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660199
CAS-Nummer: 886498-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR