3,6-Dichloro-2-fluoroaniline 98%

Lieferant: Apollo Scientific

PC302824-10G PC302824-5G PC302824-1G
APOSPC302824-10GEA 705 EUR
APOSPC302824-10G APOSPC302824-5G APOSPC302824-1G
3,6-Dichloro-2-fluoroaniline 98%
3,6-Dichloro-2-fluoroaniline
Formel: C₆H₄Cl₂FN
Molecular Weight: 180,007 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832331
CAS-Nummer: 1017778-56-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR