2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific

Synonyme: (2,3-Dichloro-6-(trifluoromethyl)phenyl)methanol

PC302906-10G PC302906-5G
APOSPC302906-10GEA 405 EUR
APOSPC302906-10G APOSPC302906-5G
2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol
2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅Cl₂F₃O
Molecular Weight: 245,026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660274
CAS-Nummer: 886501-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR