3,5-Difluoro-2-methoxybenzyl alcohol 98%

Lieferant: Apollo Scientific

PC302940-10G PC302940-5G
APOSPC302940-10GEA 316 EUR
APOSPC302940-10G APOSPC302940-5G
3,5-Difluoro-2-methoxybenzyl alcohol 98%
3,5-Difluoro-2-methoxybenzyl alcohol
Formel: C₈H₈F₂O₂
Molecular Weight: 174.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660332
CAS-Nummer: 131782-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR