4-Chloro-3,5-difluorobenzyl alcohol

Lieferant: Apollo Scientific

PC302982-10G PC302982-5G
APOSPC302982-10GEA 551 EUR
APOSPC302982-10G APOSPC302982-5G
4-Chloro-3,5-difluorobenzyl alcohol
4-Chloro-3,5-difluorobenzyl alcohol
Formel: C₇H₅ClF₂O
Molecular Weight: 178.564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23795324
CAS-Nummer: 1431329-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR