4-Fluoro-3,5-dimethylbenzoyl chloride

Lieferant: Apollo Scientific

PC302993-10G PC302993-5G
APOSPC302993-10GEA 328 EUR
APOSPC302993-10G APOSPC302993-5G
4-Fluoro-3,5-dimethylbenzoyl chloride
4-Fluoro-3,5-dimethylbenzoyl chloride
Formel: C₉H₈ClFO
Molecular Weight: 186,611 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660265
CAS-Nummer: 886501-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR