4-Fluoro-3,5-dimethylbenzyl alcohol 98%
Lieferant: Apollo Scientific
Synonyme:
(4-Fluoro-3,5-dimethylphenyl)methanol
PC302994-10G
PC302994-5G
APOSPC302994-10GEA
314
EUR
APOSPC302994-10G
APOSPC302994-5G
4-Fluoro-3,5-dimethylbenzyl alcohol 98%
4-Fluoro-3,5-dimethylbenzyl alcohol
Formel:
C₉H₁₁FO Molecular Weight: 154,181 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660267 CAS-Nummer: 886501-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...