5-Fluoro-2-methoxybenzoyl chloride

Lieferant: Apollo Scientific
Gefahr

PC303006-1G PC303006-5G
APOSPC303006-1GEA 98.6 EUR
APOSPC303006-1G APOSPC303006-5G
5-Fluoro-2-methoxybenzoyl chloride
5-Fluoro-2-methoxybenzoyl chloride
Formel: C₈H₆ClFO₂
Molecular Weight: 188.583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832309
CAS-Nummer: 704-03-0
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR