3-Butoxy-6-chloro-2-fluorobenzoic acid 97%

Lieferant: Apollo Scientific

PC303055-1G PC303055-5G
APOSPC303055-1GEA 203 EUR
APOSPC303055-1G APOSPC303055-5G
3-Butoxy-6-chloro-2-fluorobenzoic acid 97%
3-Butoxy-6-chloro-2-fluorobenzoic acid
Formel: C₁₁H₁₂ClFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27986836
CAS-Nummer: 1706435-00-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR