4-Fluoro-3,5-dimethylbenzamide 97%

Lieferant: Apollo Scientific
Achtung

PC303088-5G PC303088-1G
APOSPC303088-5GEA 203 EUR
APOSPC303088-5G APOSPC303088-1G
4-Fluoro-3,5-dimethylbenzamide 97%
4-Fluoro-3,5-dimethylbenzamide
Formel: C₉H₁₀FNO
Molecular Weight: 167.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660266
CAS-Nummer: 886501-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR