2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl alcohol 98%
Lieferant: Apollo Scientific
PC303156-1G
PC303156-250MG
APOSPC303156-1GEA
507
EUR
APOSPC303156-1G
APOSPC303156-250MG
2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl alcohol 98%
2-Fluoro-5-methyl-3-(trifluoromethyl)benzyl alcohol
Formel:
C₉H₈F₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054187 CAS-Nummer: 1706457-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...