3,4-Dichloro-5-(trifluoromethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific

PC303174-1G PC303174-250MG
APOSPC303174-1GEA 509 EUR
APOSPC303174-1G APOSPC303174-250MG
3,4-Dichloro-5-(trifluoromethyl)benzyl alcohol 97%
3,4-Dichloro-5-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅Cl₂F₃O
Molecular Weight: 245.026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28103365
CAS-Nummer: 1824048-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR