2,4,6-Trifluoro-3-methylbenzoic acid 97%

Lieferant: Apollo Scientific

PC303201-1G PC303201-250MG PC303201-5G
APOSPC303201-1GEA 866 EUR
APOSPC303201-1G APOSPC303201-250MG APOSPC303201-5G
2,4,6-Trifluoro-3-methylbenzoic acid 97%
2,4,6-Trifluoro-3-methylbenzoic acid
Formel: C₈H₅F₃O₂
Molecular Weight: 190,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27986840
CAS-Nummer: 1706435-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR