2,6-Dichloro-4-(pentafluorosulphur)benzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
PC303202-1G
PC303202-250MG
APOSPC303202-1GEA
868
EUR
APOSPC303202-1G
APOSPC303202-250MG
2,6-Dichloro-4-(pentafluorosulphur)benzenesulphonyl chloride 97%
2,6-Dichloro-4-(pentafluorosulphur)benzenesulphonyl chloride
Formel:
C₆H₂Cl₃F₅O₂S₂ Molecular Weight: 371.56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054216 CAS-Nummer: 1706430-41-2 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...