2,3-Difluoro-5-methoxy-4-methylbenzyl alcohol 98%
Lieferant: Apollo Scientific
Synonyme:
(2,3-Difluoro-5-methoxy-4-methylphenyl)methanol
PC303236-1G
PC303236-250MG
APOSPC303236-1GEA
911
EUR
APOSPC303236-1G
APOSPC303236-250MG
2,3-Difluoro-5-methoxy-4-methylbenzyl alcohol 98%
2,3-Difluoro-5-methoxy-4-methylbenzyl alcohol
Formel:
C₉H₁₀F₂O₂ Molecular Weight: 188.171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27986850 CAS-Nummer: 1706461-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...