3,4-Dichloro-5-(trifluoromethoxy)phenylacetic acid 97%
Lieferant: Apollo Scientific
PC303241-1G
PC303241-250MG
APOSPC303241-1GEA
990
EUR
APOSPC303241-1G
APOSPC303241-250MG
3,4-Dichloro-5-(trifluoromethoxy)phenylacetic acid 97%
3,4-Dichloro-5-(trifluoromethoxy)phenylacetic acid
Formel:
C₉H₅Cl₂F₃O₃ Molecular Weight: 289,035 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054209 CAS-Nummer: 1706430-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...