6-Chloro-2-fluoro-3-methoxybenzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
PC303252-1G
PC303252-250MG
APOSPC303252-1GEA
1020
EUR
APOSPC303252-1G
APOSPC303252-250MG
6-Chloro-2-fluoro-3-methoxybenzenesulphonyl chloride 97%
6-Chloro-2-fluoro-3-methoxybenzenesulphonyl chloride
Formel:
C₇H₅Cl₂FO₃S Molecular Weight: 259,082 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054220 CAS-Nummer: 1706458-71-0 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...