4-Amino-2-fluorosulphur pentafluoride 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-4-(pentafluorosulphur)aniline
PC303266-1G
PC303266-250MG
APOSPC303266-1GEA
1840
EUR
APOSPC303266-1G
APOSPC303266-250MG
4-Amino-2-fluorosulphur pentafluoride 98%
4-Amino-2-fluorosulphur pentafluoride
Formel:
C₆H₅F₆NS Molecular Weight: 237,166 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054193 CAS-Nummer: 864230-06-8 UN: 2811 ADR: 6.1,II |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...