6-Chloro-2,4-difluoro-3-methylphenylacetic acid 97%

Lieferant: Apollo Scientific

PC303283-1G PC303283-250MG
APOSPC303283-1GEA 1300 EUR
APOSPC303283-1G APOSPC303283-250MG
6-Chloro-2,4-difluoro-3-methylphenylacetic acid 97%
6-Chloro-2,4-difluoro-3-methylphenylacetic acid
Formel: C₉H₇ClF₂O₂
Molecular Weight: 220.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29038431
CAS-Nummer: 1858250-10-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR