6-Ethoxy-2,3,4-trifluorobenzyl alcohol 97%

Lieferant: Apollo Scientific

PC303289-1G PC303289-250MG
APOSPC303289-1GEA 522 EUR
APOSPC303289-1G APOSPC303289-250MG
6-Ethoxy-2,3,4-trifluorobenzyl alcohol 97%
6-Ethoxy-2,3,4-trifluorobenzyl alcohol
Formel: C₉H₉F₃O₂
Molecular Weight: 206.162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29277473
CAS-Nummer: 1980062-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR