6-Ethoxy-2,3,4-trifluorophenylacetonitrile 97%
Lieferant: Apollo Scientific
PC303291-1G
PC303291-250MG
APOSPC303291-1GEA
687
EUR
APOSPC303291-1G
APOSPC303291-250MG
6-Ethoxy-2,3,4-trifluorophenylacetonitrile 97%
6-Ethoxy-2,3,4-trifluorophenylacetonitrile
Formel:
C₁₀H₈F₃NO Molecular Weight: 215.172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29277474 CAS-Nummer: 1980053-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...