6-Ethoxy-2,3,4-trifluorophenylacetic acid 97%

Lieferant: Apollo Scientific

PC303292-1G PC303292-250MG
APOSPC303292-1GEA 800 EUR
APOSPC303292-1G APOSPC303292-250MG
6-Ethoxy-2,3,4-trifluorophenylacetic acid 97%
6-Ethoxy-2,3,4-trifluorophenylacetic acid
Formel: C₁₀H₉F₃O₃
Molecular Weight: 234.172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29277478
CAS-Nummer: 1980062-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR