3-Bromo-4-methoxy-5-(trifluoromethyl)benzenesulphonamide 97%

Lieferant: Apollo Scientific

PC303293-1G PC303293-250MG
APOSPC303293-1GEA 1390 EUR
APOSPC303293-1G APOSPC303293-250MG
3-Bromo-4-methoxy-5-(trifluoromethyl)benzenesulphonamide 97%
3-Bromo-4-methoxy-5-(trifluoromethyl)benzenesulphonamide
Formel: C₈H₇BrF₃NO₃S
Molecular Weight: 334,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29277494
CAS-Nummer: 1980053-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR