2,4-Bis(trifluoromethyl)benzenesulphonamide 97%

Lieferant: Apollo Scientific

PC303294-1G PC303294-250MG
APOSPC303294-1GEA 870 EUR
APOSPC303294-1G APOSPC303294-250MG
2,4-Bis(trifluoromethyl)benzenesulphonamide 97%
2,4-Bis(trifluoromethyl)benzenesulphonamide
Formel: C₈H₅F₆NO₂S
Molecular Weight: 293.186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29277491
CAS-Nummer: 146533-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR