6-Methoxy-2,3,4-trifluorobenzaldehyde 97%

Lieferant: Apollo Scientific

PC303298-1G PC303298-5G
APOSPC303298-1GEA 348 EUR
APOSPC303298-1G APOSPC303298-5G
6-Methoxy-2,3,4-trifluorobenzaldehyde 97%
6-Methoxy-2,3,4-trifluorobenzaldehyde
Formel: C₈H₅F₃O₂
Molecular Weight: 190.119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28801325
CAS-Nummer: 1785350-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR