2,4-Difluoro-5-methylphenylacetic acid 98%

Lieferant: Apollo Scientific

PC303305-1G PC303305-5G
APOSPC303305-1GEA 347 EUR
APOSPC303305-1G APOSPC303305-5G
2,4-Difluoro-5-methylphenylacetic acid 98%
2,4-Difluoro-5-methylphenylacetic acid
Formel: C₉H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20656599
CAS-Nummer: 367955-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR