3,6-Difluoro-2-methoxyphenylacetic acid 98%

Lieferant: Apollo Scientific

PC303317-5G PC303317-1G
APOSPC303317-5GEA 956 EUR
APOSPC303317-5G APOSPC303317-1G
3,6-Difluoro-2-methoxyphenylacetic acid 98%
3,6-Difluoro-2-methoxyphenylacetic acid
Formel: C₉H₈F₂O₃
Molecular Weight: 202,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18395260
CAS-Nummer: 1261675-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR