4-Chloro-2-fluoro-5-methylbenzyl alcohol 97%

Lieferant: Apollo Scientific

PC303321-1G PC303321-5G
APOSPC303321-1GEA 289 EUR
APOSPC303321-1G APOSPC303321-5G
4-Chloro-2-fluoro-5-methylbenzyl alcohol 97%
4-Chloro-2-fluoro-5-methylbenzyl alcohol
Formel: C₈H₈ClFO
Molecular Weight: 174.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28751970
CAS-Nummer: 1805042-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR