4-Chloro-2-fluoro-5-methylbenzyl bromide 97%

Lieferant: Apollo Scientific

PC303322-1G PC303322-5G
APOSPC303322-1GEA 287 EUR
APOSPC303322-1G APOSPC303322-5G
4-Chloro-2-fluoro-5-methylbenzyl bromide 97%
4-Chloro-2-fluoro-5-methylbenzyl bromide
Formel: C₈H₇BrClF
Molecular Weight: 237,497 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28751990
CAS-Nummer: 1805042-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR