4-Chloro-2-fluoro-5-methylphenylacetic acid 97%

Lieferant: Apollo Scientific

PC303324-1G PC303324-5G
APOSPC303324-1GEA 316 EUR
APOSPC303324-1G APOSPC303324-5G
4-Chloro-2-fluoro-5-methylphenylacetic acid 97%
4-Chloro-2-fluoro-5-methylphenylacetic acid
Formel: C₉H₈ClFO₂
Molecular Weight: 202,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28752149
CAS-Nummer: 1805227-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR