3,5-Bis(trifluoromethyl)-α-methylbenzyl bromide 98%

Lieferant: Apollo Scientific

PC303333-5G PC303333-1G
APOSPC303333-5GEA 796 EUR
APOSPC303333-5G APOSPC303333-1G
3,5-Bis(trifluoromethyl)-α-methylbenzyl bromide 98%
3,5-Bis(trifluoromethyl)-α-methylbenzyl bromide
Formel: C₁₀H₇BrF₆
Molecular Weight: 321,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18762083
CAS-Nummer: 160376-84-1
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR