2-Chloro-4-fluoro-5-methylbenzyl bromide 97%

Lieferant: Apollo Scientific

PC303339-1G PC303339-5G
APOSPC303339-1GEA 299 EUR
APOSPC303339-1G APOSPC303339-5G
2-Chloro-4-fluoro-5-methylbenzyl bromide 97%
2-Chloro-4-fluoro-5-methylbenzyl bromide
Formel: C₈H₇BrClF
Molecular Weight: 237,497 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24034366
CAS-Nummer: 1510609-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR