2-Bromo-5-chloro-4-(trifluoromethoxy)aniline 97%

Lieferant: Apollo Scientific

PC303350-5G PC303350-1G
APOSPC303350-5GEA 788 EUR
APOSPC303350-5G APOSPC303350-1G
2-Bromo-5-chloro-4-(trifluoromethoxy)aniline 97%
2-Bromo-5-chloro-4-(trifluoromethoxy)aniline
Formel: C₇H₄BrClF₃NO
Molecular Weight: 290,465 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28734569
CAS-Nummer: 868406-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR