5-Chloro-2,4-difluorobenzyl alcohol 98%

Lieferant: Apollo Scientific

PC303354-5G PC303354-1G
APOSPC303354-5GEA 512 EUR
APOSPC303354-5G APOSPC303354-1G
5-Chloro-2,4-difluorobenzyl alcohol 98%
5-Chloro-2,4-difluorobenzyl alcohol
Formel: C₇H₅ClF₂O
Molecular Weight: 178,564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22200072
CAS-Nummer: 915409-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR