5-Chloro-2,4-difluorobenzylamine

Lieferant: Apollo Scientific

Synonyme: (5-Chloro-2,4-difluorophenyl)methanamine

PC303359-1G PC303359-5G
APOSPC303359-1GEA 77 EUR
APOSPC303359-1G APOSPC303359-5G
5-Chloro-2,4-difluorobenzylamine
5-Chloro-2,4-difluorobenzylamine
Formel: C₇H₆ClF₂N
Molecular Weight: 177,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458696
CAS-Nummer: 924818-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR