5-Chloro-2,4-difluorobenzamide 97%

Lieferant: Apollo Scientific

PC303361-5G PC303361-1G
APOSPC303361-5GEA 397 EUR
APOSPC303361-5G APOSPC303361-1G
5-Chloro-2,4-difluorobenzamide 97%
5-Chloro-2,4-difluorobenzamide
Formel: C₇H₄ClF₂NO
Molecular Weight: 191,563 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28750484
CAS-Nummer: 1805526-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR