4,6-Dichloro-2-fluoro-3-methoxybenzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

PC303367-5G PC303367-1G
APOSPC303367-5GEA 940 EUR
APOSPC303367-5G APOSPC303367-1G
4,6-Dichloro-2-fluoro-3-methoxybenzaldehyde 98%
4,6-Dichloro-2-fluoro-3-methoxybenzaldehyde
Formel: C₈H₅Cl₂FO₂
Molecular Weight: 223,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30726936
CAS-Nummer: 2149599-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR