4,6-Dichloro-2-fluoro-3-methoxybenzyl alcohol 97%

Lieferant: Apollo Scientific

PC303368-1G PC303368-5G
APOSPC303368-1GEA 277 EUR
APOSPC303368-1G APOSPC303368-5G
4,6-Dichloro-2-fluoro-3-methoxybenzyl alcohol 97%
4,6-Dichloro-2-fluoro-3-methoxybenzyl alcohol
Formel: C₈H₇Cl₂FO₂
Molecular Weight: 225.044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30726937
CAS-Nummer: 2149598-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR