4,6-Dichloro-2-fluoro-3-methoxyphenylacetic acid

Lieferant: Apollo Scientific

PC303371-1G PC303371-5G
APOSPC303371-1GEA 339 EUR
APOSPC303371-1G APOSPC303371-5G
4,6-Dichloro-2-fluoro-3-methoxyphenylacetic acid
4,6-Dichloro-2-fluoro-3-methoxyphenylacetic acid
Formel: C₉H₇Cl₂FO₃
Molecular Weight: 253,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30726939
CAS-Nummer: 2149589-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR