4-Bromo-2-chloro-6-(trifluoromethyl)benzenesulphonyl chloride 97%

Lieferant: Apollo Scientific

PC303390-1G PC303390-250MG
APOSPC303390-1GEA 894 EUR
APOSPC303390-1G APOSPC303390-250MG
4-Bromo-2-chloro-6-(trifluoromethyl)benzenesulphonyl chloride 97%
4-Bromo-2-chloro-6-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂BrCl₂F₃O₂S
Molecular Weight: 357.96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31916398
CAS-Nummer: 2384019-85-4
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR