6-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303397-1G PC303397-5G
APOSPC303397-1GEA 60.9 EUR
APOSPC303397-1G APOSPC303397-5G
6-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%
6-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid
Formel: C₉H₆F₄O₂
Molecular Weight: 222,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640056
CAS-Nummer: 2383927-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR