6-Chloro-2,4-difluoro-3-methylbenzylamine 98%

Lieferant: Apollo Scientific

PC303403-1G PC303403-250MG
APOSPC303403-1GEA 1280 EUR
APOSPC303403-1G APOSPC303403-250MG
6-Chloro-2,4-difluoro-3-methylbenzylamine 98%
6-Chloro-2,4-difluoro-3-methylbenzylamine
Formel: C₈H₈ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD32640057
CAS-Nummer: 2415751-58-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR