4-(3,5-Difluorophenoxy)-3-bromoaniline 98%

Lieferant: Apollo Scientific

PC303421-10G PC303421-1G PC303421-5G
APOSPC303421-10GEA 416 EUR
APOSPC303421-10G APOSPC303421-1G APOSPC303421-5G
4-(3,5-Difluorophenoxy)-3-bromoaniline 98%
4-(3,5-Difluorophenoxy)-3-bromoaniline
Formel: C₁₂H₈BrF₂NO
Molecular Weight: 300,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17121998
CAS-Nummer: 1483390-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR